Assay Details
| Gene Symbol: CCR10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194964.1 | NP_001181893.1 | 2 | 823 | 80 | |
| GenBank mRNA | EU684266.1 | - | 1 | 781 | 80 | |
| GU936965.1 | - | 2 | 823 | 80 | ||
More Information
Set Membership: |
|

CCR10
Genomic Map