Assay Details
| Gene Symbol: JUN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077827.1 | NP_001071295.1 | 2 | 1721 | 91 | |
| GenBank mRNA | BC123753.1 | - | 2 | 1721 | 91 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: JUN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077827.1 | NP_001071295.1 | 2 | 1721 | 91 | |
| GenBank mRNA | BC123753.1 | - | 2 | 1721 | 91 | |
Set Membership: |
|