Assay Details
| Gene Symbol: MT1E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114857.2 | NP_001108329.1 | 324 | 93 | ||
| GenBank mRNA | BC118366.1 | - | 325 | 93 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MT1E| Gene Symbol: MT1E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114857.2 | NP_001108329.1 | 324 | 93 | ||
| GenBank mRNA | BC118366.1 | - | 325 | 93 | ||
Set Membership: |
|