Assay Details
| Gene Symbol: ENSA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001314005.1 | NP_001300934.1 | 2 - 3 | 314 | 70 | |
| NM_174050.2 | NP_776475.2 | 2 - 3 | 314 | 70 | ||
| GenBank mRNA | AJ005986.1 | - | 2 - 3 | 182 | 70 | |
| BC112682.1 | - | 2 - 3 | 311 | 70 | ||
| EV629449.1 | - | 2 - 3 | 317 | 70 | ||
| EV691157.1 | - | 2 - 3 | 241 | 70 | ||
More Information
Set Membership: |
|

ENSA
Genomic Map