Assay Details
| Gene Symbol: MIR15B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_031363.1 | 1 | 49 | 83 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA mir-16b | MIR16B |
MIRN16, bta-mir-16, bta-mir-16b, mir-16b |
-51.0 | Sense | Chr. 1 - 107073622 - 107073714 on NCBI Build ARS-UCD1.2 |
| Downstream | intraflagellar transport 80 | IFT80 | 5.0kb | Anti-sense | Chr. 1 - 107078906 - 107210812 on NCBI Build ARS-UCD1.2 |
More Information
Set Membership: |
|

MIR15B
Genomic Map