Assay Details
Gene Symbol: PPARG-TSEN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_003096.1 | 8 - 9 | 1430 | 85 | ||
GenBank mRNA | DQ256759.1 | - | 8 - 9 | 1430 | 85 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | makorin ring finger protein 2 | MKRN2 | -73.6kb | Sense | Chr. 22 - 56543251 - 56581386 on NCBI Build ARS-UCD1.2 | |
Downstream | microRNA mir-2373 | MIR2373 | bta-mir-2373 | 104.2kb | Anti-sense | Chr. 22 - 56833146 - 56833223 on NCBI Build ARS-UCD1.2 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
