Assay Details
| Gene Symbol: MIR129-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_030802.1 | 1 | 28 | 60 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | staphylococcal nuclease and tudor domain containing 1 | SND1 | -111.8kb | Sense | Chr. 4 - 91873942 - 92296112 on NCBI Build ARS-UCD1.2 | |
| Downstream | leptin | LEP | ob | 28.8kb | Sense | Chr. 4 - 92436837 - 92453660 on NCBI Build ARS-UCD1.2 |
More Information
Set Membership: |
|

MIR129-1
Genomic Map