Assay Details
Gene Symbol: MIR2453 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_030811.1 | 1 | 33 | 65 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | coiled-coil domain containing 96 | CCDC96 | -10.0kb | Anti-sense | Chr. 6 - 113508435 - 113511325 on NCBI Build ARS-UCD1.2 | |
Downstream |
GrpE like 1, mitochondrial |
GRPEL1 | 7.6kb | Anti-sense | Chr. 6 - 113529004 - 113543058 on NCBI Build ARS-UCD1.2 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
