Assay Details
Gene Symbol: MIR708 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_030987.1 | 1 | 59 | 80 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | family with sequence similarity 181 member B | FAM181B | -3.7mb | Sense | Chr. 29 - 12925471 - 12927326 on NCBI Build ARS-UCD1.2 | |
Downstream | JY-1 | LOC100126230 | 810.2kb | Sense | Chr. 29 - 17406353 - 17422383 on NCBI Build ARS-UCD1.2 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
