Assay Details
Gene Symbol: MIR2423 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_030878.1 | 1 | 38 | 65 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | coiled-coil-helix-coiled-coil-helix domain containing 3 | CHCHD3 | -456.1kb | Anti-sense | Chr. 4 - 96468819 - 96761374 on NCBI Build ARS-UCD1.2 | |
Downstream | leucine rich repeats and guanylate kinase domain containing | LRGUK | 572.4kb | Sense | Chr. 4 - 97789905 - 97893782 on NCBI Build ARS-UCD1.2 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
