Assay Details
Gene Symbol: MIR2433 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_030903.1 | 1 | 36 | 56 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | coenzyme Q10A | COQ10A | -6.5kb | Sense | Chr. 5 - 57065731 - 57069113 on NCBI Build ARS-UCD1.2 | |
Downstream | solute carrier family 39 member 5 | SLC39A5 | 16.0kb | Sense | Chr. 5 - 57091700 - 57097291 on NCBI Build ARS-UCD1.2 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
