Assay Details
Gene Symbol: MIR2430 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_030904.1 | 1 | 32 | 66 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | DNA damage inducible transcript 3 | DDIT3 |
CHOP, CHOP-10 |
-4.0kb | Anti-sense | Chr. 5 - 55956303 - 55960513 on NCBI Build ARS-UCD1.2 |
Downstream | GLI family zinc finger 1 | GLI1 | GLI | 28.1kb | Sense | Chr. 5 - 55992681 - 56003151 on NCBI Build ARS-UCD1.2 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
