Assay Details
| Gene Symbol: MIR205 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_030909.1 | 1 | 33 | 66 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | calcium/calmodulin dependent protein kinase IG | CAMK1G | -156.5kb | Sense | Chr. 16 - 73713231 - 73744186 on NCBI Build ARS-UCD1.2 | |
| Downstream | plexin A2 | PLXNA2 | 1.2mb | Anti-sense | Chr. 16 - 75115600 - 75353862 on NCBI Build ARS-UCD1.2 |
More Information
Set Membership: |
|

MIR205
Genomic Map