Assay Details
| Gene Symbol: MIR412 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_031065.1 | 1 | 49 | 82 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA mir-409b | MIR409B |
MIR3581, bta-mir-3581, bta-mir-409b |
-64.0 | Anti-sense | Chr. 21 - 65953893 - 65953971 on NCBI Build ARS-UCD1.2 |
| Downstream | microRNA mir-369 | MIR369 |
MIRN369, bta-mir-369 |
64.0 | Sense | Chr. 21 - 65954188 - 65954256 on NCBI Build ARS-UCD1.2 |
More Information
Set Membership: |
|

MIR412
Genomic Map