Assay Details
Gene Symbol: MIR2294 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_031089.1 | 1 | 34 | 58 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | SMYD family member 5 | SMYD5 | -95.2kb | Anti-sense | Chr. 11 - 11220296 - 11232167 on NCBI Build ARS-UCD1.2 | |
Downstream | sideroflexin 5 | SFXN5 | 44.1kb | Sense | Chr. 11 - 11371501 - 11498288 on NCBI Build ARS-UCD1.2 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
