Assay Details
| Gene Symbol: MIR2314 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_031163.1 | 1 | 30 | 60 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | aryl hydrocarbon receptor nuclear translocator like | ARNTL | -407.9kb | Sense | Chr. 15 - 39286453 - 39394465 on NCBI Build ARS-UCD1.2 | |
| Downstream | parvin alpha | PARVA | 365.6kb | Sense | Chr. 15 - 40168037 - 40347128 on NCBI Build ARS-UCD1.2 |
More Information
Set Membership: |
|

MIR2314
Genomic Map