Assay Details
| Gene Symbol: MIR2395 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_031171.1 | 1 | 36 | 57 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | chromosome 26 C10orf82 homolog | C26H10orf82 | -26.0kb | Sense | Chr. 26 - 37058422 - 37066152 on NCBI Build ARS-UCD1.2 | |
| Downstream | enolase 4 | ENO4 | 145.5kb | Anti-sense | Chr. 26 - 37237721 - 37268633 on NCBI Build ARS-UCD1.2 |
More Information
Set Membership: |
|

MIR2395
Genomic Map