Assay Details
Gene Symbol: MIR187 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_031180.1 | 1 | 54 | 80 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | chromosome 24 C18orf21 homolog | C24H18orf21 | C18orf21 | -73.0kb | Anti-sense | Chr. 24 - 21138186 - 21146243 on NCBI Build ARS-UCD1.2 |
Downstream | polypeptide N-acetylgalactosaminyltransferase 1 | GALNT1 | galNAc-T1 | 155.3kb | Anti-sense | Chr. 24 - 21374710 - 21455432 on NCBI Build ARS-UCD1.2 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
