Assay Details
| Gene Symbol: MIR33A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_031206.1 | 1 | 33 | 67 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | sterol regulatory element binding transcription factor 2 | SREBF2 | Sense | Chr. 5 - 112832407 - 112890177 on NCBI Build ARS-UCD1.2 | ||
| Downstream | TNF receptor superfamily member 13C | TNFRSF13C | 21.2kb | Anti-sense | Chr. 5 - 112905849 - 112907879 on NCBI Build ARS-UCD1.2 |
More Information
Set Membership: |
|

MIR33A
Genomic Map