Assay Details
Gene Symbol: MIR2462 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_031212.1 | 1 | 24 | 66 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | methionine adenosyltransferase 2B | MAT2B | -5.2mb | Sense | Chr. 7 - 75184589 - 75202950 on NCBI Build ARS-UCD1.2 | |
Downstream | arginyl-tRNA synthetase 1 | RARS1 |
ArgRS, RARS |
147.1kb | Sense | Chr. 7 - 80503748 - 80532990 on NCBI Build ARS-UCD1.2 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
