Assay Details
Gene Symbol: MIR151 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_031270.1 | 1 | 30 | 63 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | protein tyrosine phosphatase 4A3 | PTP4A3 | -416.6kb | Anti-sense | Chr. 14 - 2553478 - 2585771 on NCBI Build ARS-UCD1.2 | |
Downstream | argonaute RISC catalytic component 2 | AGO2 | EIF2C2 | 98.4kb | Sense | Chr. 14 - 3100810 - 3138228 on NCBI Build ARS-UCD1.2 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
