Assay Details
Gene Symbol: MIR2485 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_031276.1 | 1 | 32 | 58 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | renin binding protein | RENBP |
AGE, RnBP |
-18.6kb | Sense | Chr. X - 37149900 - 37157509 on NCBI Build ARS-UCD1.2 |
Downstream | transmembrane protein 187 | TMEM187 | 7.1kb | Anti-sense | Chr. X - 37183241 - 37188624 on NCBI Build ARS-UCD1.2 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
