Assay Details
Gene Symbol: MIR2372 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_031299.1 | 1 | 38 | 68 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | thyrotropin releasing hormone | TRH | -168.1kb | Anti-sense | Chr. 22 - 55886890 - 55889851 on NCBI Build ARS-UCD1.2 | |
Downstream | H1.8 linker histone | H1-8 | H1FOO | 129.6kb | Anti-sense | Chr. 22 - 56187659 - 56223207 on NCBI Build ARS-UCD1.2 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
