Assay Details
Gene Symbol: MIR449D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_031303.1 | 1 | 41 | 65 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | developing brain homeobox 1 | DBX1 | -200.4kb | Anti-sense | Chr. 29 - 24757938 - 24762782 on NCBI Build ARS-UCD1.2 | |
Downstream | E2F transcription factor 8 | E2F8 | E2F-8 | 735.9kb | Anti-sense | Chr. 29 - 25699179 - 25717572 on NCBI Build ARS-UCD1.2 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
