Assay Details
| Gene Symbol: MIR2334 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_030855.1 | 1 | 39 | 69 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ankyrin repeat domain 13B | ANKRD13B | -7.6kb | Anti-sense | Chr. 19 - 20848016 - 20864213 on NCBI Build ARS-UCD1.2 | |
| Downstream | nuclear speckle splicing regulatory protein 1 | NSRP1 |
CCDC55, NSrp70 |
365.8kb | Anti-sense | Chr. 19 - 21237745 - 21283421 on NCBI Build ARS-UCD1.2 |
More Information
Set Membership: |
|

MIR2334
Genomic Map