Assay Details
Gene Symbol: MIR562 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_031333.1 | 1 | 44 | 74 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | bifunctional apoptosis regulator | BFAR | -117.9kb | Anti-sense | Chr. 25 - 13548018 - 13579860 on NCBI Build ARS-UCD1.2 | |
Downstream | N-terminal asparagine amidase | NTAN1 | 18.8kb | Anti-sense | Chr. 25 - 13716625 - 13731513 on NCBI Build ARS-UCD1.2 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
