Assay Details
Gene Symbol: MIR2477 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_030929.1 | 1 | 35 | 65 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | HD domain containing 2 | HDDC2 | Sense | Chr. 9 - 25960234 - 25979949 on NCBI Build ARS-UCD1.2 | ||
Downstream | TPD52 like 1 | TPD52L1 | 7.2kb | Anti-sense | Chr. 9 - 25985760 - 26078941 on NCBI Build ARS-UCD1.2 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
