Assay Details
Gene Symbol: MIR592 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_031341.1 | 1 | 54 | 73 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | glutamate metabotropic receptor 8 | GRM8 | Sense | Chr. 4 - 90671110 - 91521030 on NCBI Build ARS-UCD1.2 | ||
Downstream | zinc finger protein 800 | ZNF800 | 294.2kb | Sense | Chr. 4 - 91614929 - 91639109 on NCBI Build ARS-UCD1.2 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
