Assay Details
Gene Symbol: MIR200A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_031343.1 | 1 | 45 | 64 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | microRNA mir-429 | MIR429 |
bta-mir-429, mir-429 |
-1.2kb | Sense | Chr. 16 - 51321445 - 51321528 on NCBI Build ARS-UCD1.2 |
Downstream | microRNA mir-200b | MIR200B |
MIRN200B, bta-mir-200b, mir-200b |
583.0 | Sense | Chr. 16 - 51323348 - 51323406 on NCBI Build ARS-UCD1.2 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
