Assay Details
| Gene Symbol: SNHG4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_038087.1 | 2 - 3 | 136 | 59 | ||
| GenBank mRNA | DT720536.1 | - | 2 - 3 | 105 | 59 | |
| DV890667.1 | - | 2 - 3 | 136 | 59 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | SIL1 nucleotide exchange factor | SIL1 | -62.5kb | Anti-sense | Chr. 7 - 50263858 - 50520143 on NCBI Build ARS-UCD1.2 | |
| Downstream | matrin 3 | MATR3 | 9.4kb | Sense | Chr. 7 - 50593615 - 50620420 on NCBI Build ARS-UCD1.2 |
More Information
Set Membership: |
|

SNHG4
Genomic Map