Assay Details
Gene Symbol: LOC100196901 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_038107.2 | 5 - 6 | 435 | 87 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | ST6 N-acetylgalactosaminide alpha-2,6-sialyltransferase 2 | ST6GALNAC2 | -54.0 | Anti-sense | Chr. 19 - 55174159 - 55194286 on NCBI Build ARS-UCD1.2 | |
Downstream | cytoglobin | CYGB | GLNF | 22.3kb | Anti-sense | Chr. 19 - 55218139 - 55227970 on NCBI Build ARS-UCD1.2 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
