Assay Details
Gene Symbol: MGC148318 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_038117.1 | 2 - 3 | 398 | 109 | ||
GenBank mRNA | BC149268.1 | - | 2 - 3 | 139 | 109 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | small nuclear RNA activating complex polypeptide 1 | SNAPC1 | -332.9kb | Sense | Chr. 10 - 73888973 - 73918569 on NCBI Build ARS-UCD1.2 | |
Downstream | potassium voltage-gated channel subfamily H member 5 | KCNH5 | 677.9kb | Anti-sense | Chr. 10 - 74940464 - 75324314 on NCBI Build ARS-UCD1.2 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC149268.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
