Assay Details
| Gene Symbol: MGC148318 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_038117.1 | 3 - 4 | 517 | 97 | ||
| GenBank mRNA | BC149268.1 | - | 3 - 4 | 258 | 97 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | small nuclear RNA activating complex polypeptide 1 | SNAPC1 | -343.9kb | Sense | Chr. 10 - 73888973 - 73918569 on NCBI Build ARS-UCD1.2 | |
| Downstream | potassium voltage-gated channel subfamily H member 5 | KCNH5 | 674.9kb | Anti-sense | Chr. 10 - 74940464 - 75324314 on NCBI Build ARS-UCD1.2 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC149268.1 transcripts.
Set Membership: |
|

MGC148318
Genomic Map