Assay Details
Gene Symbol: MIR6520 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_107757.1 | 1 | 42 | 82 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | polypeptide N-acetylgalactosaminyltransferase 9 | GALNT9 | WBSCR17 | -122.9kb | Anti-sense | Chr. 17 - 44853960 - 44973834 on NCBI Build ARS-UCD1.2 |
Downstream | pseudouridine synthase 1 | PUS1 | 65.8kb | Sense | Chr. 17 - 45162664 - 45173111 on NCBI Build ARS-UCD1.2 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
