Assay Details
Gene Symbol: MIR2284Y-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_107778.1 | 1 | 49 | 72 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | paired box 3 | PAX3 | Anti-sense | Chr. 2 - 110369553 - 110471285 on NCBI Build ARS-UCD1.2 | ||
Downstream | phenylalanyl-tRNA synthetase subunit beta | FARSB | 368.0kb | Anti-sense | Chr. 2 - 110746220 - 110820162 on NCBI Build ARS-UCD1.2 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
