Assay Details
| Gene Symbol: MIR503 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_107789.1 | 1 | 41 | 64 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA mir-542 | MIR542 | bta-mir-542 | -4.9kb | Sense | Chr. X - 18237439 - 18237535 on NCBI Build ARS-UCD1.2 |
| Downstream | microRNA mir-424 | MIR424 |
bta-mir-424, mir-424 |
262.0 | Sense | Chr. X - 18242745 - 18242840 on NCBI Build ARS-UCD1.2 |
More Information
Set Membership: |
|

MIR503
Genomic Map