Assay Details
Gene Symbol: MIR219-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_107849.1 | 1 | 34 | 59 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | hydroxysteroid 17-beta dehydrogenase 8 | HSD17B8 |
FABGL, HKE6 |
-1.0kb | Sense | Chr. 23 - 7391656 - 7393772 on NCBI Build ARS-UCD1.2 |
Downstream | ring finger protein 1 | RING1 | 680.0 | Sense | Chr. 23 - 7395562 - 7399419 on NCBI Build ARS-UCD1.2 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
