Assay Details
| Gene Symbol: RNASEK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_130175.1 | 1 - 2 | 209 | 88 | ||
| GenBank mRNA | EH143926.1 | - | 1 - 2 | 163 | 88 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
arachidonate 12-lipoxygenase, 12S type |
ALOX12 | 12-LOX | -1.8kb | Sense | Chr. 19 - 26786538 - 26800876 on NCBI Build ARS-UCD1.2 |
| Downstream | chromosome 19 C17orf49 homolog | C19H17orf49 |
BAP18, C17orf49 |
1.1kb | Sense | Chr. 19 - 26804519 - 26807430 on NCBI Build ARS-UCD1.2 |
More Information
Set Membership: |
|

RNASEK
Genomic Map