Assay Details
| Gene Symbol: CLEC1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001281914.1 | NP_001268843.1 | 1 - 2 | 67 | 76 | |
| GenBank mRNA | HQ738491.1 | - | 1 - 2 | 67 | 76 | |
More Information
Set Membership: |
|

CLEC1B
Genomic Map