Assay Details
| Gene Symbol: BLMH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001303621.1 | NP_001290550.1 | 9 - 10 | 1234 | 61 | |
| GenBank mRNA | EE332757.1 | - | 4 - 5 | 488 | 61 | |
More Information
Set Membership: |
|

BLMH
Genomic Map