Assay Details
| Gene Symbol: LOC104968411 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC149909.1 | - | 18 - 19 | 2965 | 58 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC104968411| Gene Symbol: LOC104968411 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC149909.1 | - | 18 - 19 | 2965 | 58 | |
Set Membership: |
|