Assay Details
| Gene Symbol: LOC104968411 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC149909.1 | - | 7 - 8 | 1022 | 105 | |
| U63924.1 | - | 44 - 45 | 6772 | 105 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC104968411| Gene Symbol: LOC104968411 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC149909.1 | - | 7 - 8 | 1022 | 105 | |
| U63924.1 | - | 44 - 45 | 6772 | 105 | ||
Set Membership: |
|