Assay Details
| Gene Symbol: GPIHBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | KF855956.1 | - | 3 - 4 | 273 | 75 | |
| KF855957.1 | - | 3 - 4 | 312 | 75 | ||
| KJ502292.1 | - | 4 - 5 | 458 | 75 | ||
More Information
Set Membership: |
|

GPIHBP1
Genomic Map