Assay Details
| Gene Symbol: LOC511354 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JQ012926.1 | - | 1 - 2 | 35 | 101 | |
| Gene Symbol: LOC615227 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BF889951.1 | - | 1 - 2 | 61 | 101 | |
More Information
Set Membership: |
|

LOC511354
Genomic Map