Assay Details
| Gene Symbol: IL13RA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206677.2 | NP_001193606.1 | 9 - 10 | 1140 | 70 | |
| GenBank mRNA | AF074402.1 | - | 8 - 9 | 1001 | 70 | |
| EE332742.1 | - | 4 - 5 | 366 | 70 | ||
More Information
Set Membership: |
|

IL13RA1
Genomic Map