Assay Details
| Gene Symbol: nlp-40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058259.5 | NP_490661.1 | 2 - 3 | 145 | 84 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

nlp-40| Gene Symbol: nlp-40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058259.5 | NP_490661.1 | 2 - 3 | 145 | 84 | |
Set Membership: |
|