Assay Details
| Gene Symbol: pgs-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001377579.2 | NP_001364825.1 | 2 - 3 | 147 | 77 | |
| NM_058265.6 | NP_490666.2 | 4 - 5 | 546 | 77 | ||
More Information
Set Membership: |
|

pgs-1
Genomic Map