Assay Details
Gene Symbol: spe-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_058279.2 | NP_490680.1 | 5 - 6 | 1227 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
