Assay Details
| Gene Symbol: tap-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078027.6 | NP_510428.1 | 6 - 7 | 812 | 111 | |
| GenBank mRNA | AF145375.1 | - | 5 - 6 | 606 | 111 | |
More Information
Set Membership: |
|

tap-1
Genomic Map