Assay Details
Gene Symbol: rbg-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078089.5 | NP_510490.1 | 6 - 7 | 751 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
